missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)phenyl Isothiocyanate 98.0+%, TCI America™

Catalog No. T33411G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33411G 1 g
T33415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33411G Supplier TCI America Supplier No. T33411G

Chemical Identifiers

CAS 64285-95-6
Molecular Formula C8H4F3NOS
Molecular Weight (g/mol) 219.181
MDL Number MFCD00042412
InChI Key JKSZUQPHKOPVHF-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy phenyl isothiocyanate, 1-isothiocyanato-4-trifluoromethoxy benzene, benzene,1-isothiocyanato-4-trifluoromethoxy, benzene, 1-isothiocyanato-4-trifluoromethoxy, 4-trifluoromethoxy phenylisothiocyanate, 1-isothiocyanato-4-trifluoromethoxy-benzene, acmc-1b5bx, 4-trifluoromethoxyphenyl isothiocyanate, 4-trifluoromethoxyphenyl-isothiocyanate, 4-trifluoromethoxy benzenisothiocyanate
PubChem CID 2777338
IUPAC Name 1-isothiocyanato-4-(trifluoromethoxy)benzene
SMILES C1=CC(=CC=C1N=C=S)OC(F)(F)F

Specifications

Boiling Point 88°C
Quantity 1 g
UN Number 1760
Formula Weight 219.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethoxy)phenyl Isothiocyanate
TSCA No
Recommended Storage Refrigerator