missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)phenyl isothiocyanate, 97%

Catalog No. AAH6401306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6401306 5 g
AAH6401303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6401306 Supplier Thermo Scientific Chemicals Supplier No. H6401306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 64285-95-6 | C8H4F3NOS | 219.181 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 64285-95-6
Molecular Formula C8H4F3NOS
Molecular Weight (g/mol) 219.181
MDL Number MFCD00042412
InChI Key JKSZUQPHKOPVHF-UHFFFAOYSA-N
PubChem CID 2777338
IUPAC Name 1-isothiocyanato-4-(trifluoromethoxy)benzene
SMILES C1=CC(=CC=C1N=C=S)OC(F)(F)F

Specifications

Color Colorless
Density 1.35 g/mL
Boiling Point 215°C to 218°C (literature)
Flash Point 107°C (225°F)
Refractive Index 1.541
Quantity 5 g
UN Number UN1760
Beilstein 2723939
Sensitivity Moisture sensitive
Synonym 4-trifluoromethoxy phenyl isothiocyanate, 1-isothiocyanato-4-trifluoromethoxy benzene, benzene,1-isothiocyanato-4-trifluoromethoxy, benzene, 1-isothiocyanato-4-trifluoromethoxy, 4-trifluoromethoxy phenylisothiocyanate, 1-isothiocyanato-4-trifluoromethoxy-benzene, acmc-1b5bx, 4-trifluoromethoxyphenyl isothiocyanate, 4-trifluoromethoxyphenyl-isothiocyanate, 4-trifluoromethoxy benzenisothiocyanate
Formula Weight 219.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethoxy)phenyl isothiocyanate
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only