missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)phenyl Isocyanate 98.0+%, TCI America™

Supplier:  TCI America T248725G

 View more versions of this product

Catalog No. T248725G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-(Trifluoromethoxy)phenyl Isocyanate
Yellow
C8H4F3NO2
25 g
4-trifluoromethoxy phenyl isocyanate, 1-isocyanato-4-trifluoromethoxy benzene, 4-trifluoromethoxy phenylisocyanate, 4-trifluoromethoxyphenylisocyanate, benzene, 1-isocyanato-4-trifluoromethoxy, 1-isocyanato-4-trifluoromethoxy-benzene, p-trifluoromethoxy phenyl isocyanate, isocyanic acid 4-trifluoromethoxy phenyl ester, 4-trifluoromethoxy benzenisocyanate
C1=CC(=CC=C1N=C=O)OC(F)(F)F
203.12
203.12
Liquid
35037-73-1
78°C
MFCD00035702
2206
LGPKFIGMLPDYEA-UHFFFAOYSA-N
1-isocyanato-4-(trifluoromethoxy)benzene
92298
≥98.0% (GC)

Chemical Identifiers

35037-73-1
203.12
LGPKFIGMLPDYEA-UHFFFAOYSA-N
92298
C1=CC(=CC=C1N=C=O)OC(F)(F)F
C8H4F3NO2
MFCD00035702
4-trifluoromethoxy phenyl isocyanate, 1-isocyanato-4-trifluoromethoxy benzene, 4-trifluoromethoxy phenylisocyanate, 4-trifluoromethoxyphenylisocyanate, benzene, 1-isocyanato-4-trifluoromethoxy, 1-isocyanato-4-trifluoromethoxy-benzene, p-trifluoromethoxy phenyl isocyanate, isocyanic acid 4-trifluoromethoxy phenyl ester, 4-trifluoromethoxy benzenisocyanate
1-isocyanato-4-(trifluoromethoxy)benzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions