missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Trifluoromethoxy)phenyl Isocyanate 98.0+%, TCI America™
$50.33 - $139.81
Chemical Identifiers
| CAS | 35037-73-1 |
|---|---|
| Molecular Formula | C8H4F3NO2 |
| Molecular Weight (g/mol) | 203.12 |
| MDL Number | MFCD00035702 |
| InChI Key | LGPKFIGMLPDYEA-UHFFFAOYSA-N |
| Synonym | 4-trifluoromethoxy phenyl isocyanate, 1-isocyanato-4-trifluoromethoxy benzene, 4-trifluoromethoxy phenylisocyanate, 4-trifluoromethoxyphenylisocyanate, benzene, 1-isocyanato-4-trifluoromethoxy, 1-isocyanato-4-trifluoromethoxy-benzene, p-trifluoromethoxy phenyl isocyanate, isocyanic acid 4-trifluoromethoxy phenyl ester, 4-trifluoromethoxy benzenisocyanate |
| PubChem CID | 92298 |
| IUPAC Name | 1-isocyanato-4-(trifluoromethoxy)benzene |
| SMILES | C1=CC(=CC=C1N=C=O)OC(F)(F)F |
Chemical Identifiers
| 35037-73-1 | |
| 203.12 | |
| LGPKFIGMLPDYEA-UHFFFAOYSA-N | |
| 92298 | |
| C1=CC(=CC=C1N=C=O)OC(F)(F)F |
| C8H4F3NO2 | |
| MFCD00035702 | |
| 4-trifluoromethoxy phenyl isocyanate, 1-isocyanato-4-trifluoromethoxy benzene, 4-trifluoromethoxy phenylisocyanate, 4-trifluoromethoxyphenylisocyanate, benzene, 1-isocyanato-4-trifluoromethoxy, 1-isocyanato-4-trifluoromethoxy-benzene, p-trifluoromethoxy phenyl isocyanate, isocyanic acid 4-trifluoromethoxy phenyl ester, 4-trifluoromethoxy benzenisocyanate | |
| 1-isocyanato-4-(trifluoromethoxy)benzene |
Specifications
| 35037-73-1 | |
| 78°C | |
| MFCD00035702 | |
| 2206 | |
| LGPKFIGMLPDYEA-UHFFFAOYSA-N | |
| 1-isocyanato-4-(trifluoromethoxy)benzene | |
| 92298 | |
| ≥98.0% (GC) | |
| 4-(Trifluoromethoxy)phenyl Isocyanate |
| Yellow | |
| C8H4F3NO2 | |
| 5 g | |
| 4-trifluoromethoxy phenyl isocyanate, 1-isocyanato-4-trifluoromethoxy benzene, 4-trifluoromethoxy phenylisocyanate, 4-trifluoromethoxyphenylisocyanate, benzene, 1-isocyanato-4-trifluoromethoxy, 1-isocyanato-4-trifluoromethoxy-benzene, p-trifluoromethoxy phenyl isocyanate, isocyanic acid 4-trifluoromethoxy phenyl ester, 4-trifluoromethoxy benzenisocyanate | |
| C1=CC(=CC=C1N=C=O)OC(F)(F)F | |
| 203.12 | |
| 203.12 | |
| Liquid |
Safety and Handling
TSCA : Yes