missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)phenol 95.0+%, TCI America™

Catalog No. p-7136723
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13415G 5 g
T134125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T13415G Supplier TCI America Supplier No. T13415G
Only null left

Chemical Identifiers

CAS 828-27-3
Molecular Formula C7H5F3O2
Molecular Weight (g/mol) 178.11
MDL Number MFCD00040988
InChI Key WDRJNKMAZMEYOF-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy phenol, p-trifluoromethoxy phenol, phenol, 4-trifluoromethoxy, 4-hydroxytrifluoromethoxybenzene, 4-trifluoromethoxy-phenol, 4-hydroxyphenyl trifluoromethyl ether, p-trifluoromethoxyphenol
PubChem CID 70015
IUPAC Name 4-(trifluoromethoxy)phenol
SMILES OC1=CC=C(OC(F)(F)F)C=C1

Specifications

Color Yellow
Boiling Point 95°C
Quantity 5 g
Formula Weight 178.11
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethoxy)phenol