missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)phenol, 98%

Catalog No. AC306040010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC306040010 1 g Glass bottle
AC306040050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC306040010 Supplier Thermo Scientific Chemicals Supplier No. 306040010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 828-27-3 | C7H5F3O2 | 178.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 828-27-3
Molecular Formula C7H5F3O2
Molecular Weight (g/mol) 178.11
MDL Number MFCD00040988
InChI Key WDRJNKMAZMEYOF-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy phenol, p-trifluoromethoxy phenol, phenol, 4-trifluoromethoxy, 4-hydroxytrifluoromethoxybenzene, 4-trifluoromethoxy-phenol, 4-hydroxyphenyl trifluoromethyl ether, p-trifluoromethoxyphenol
PubChem CID 70015
IUPAC Name 4-(trifluoromethoxy)phenol
SMILES OC1=CC=C(OC(F)(F)F)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Brown
Density 1.3700g/mL
Boiling Point 80.0°C (15.0 mmHg)
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4460 to 1.4480
Linear Formula CF3OC6H4OH
Quantity 1 g
Specific Gravity 1.37
Formula Weight 178.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethoxy)phenol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 212-583-

RUO – Research Use Only