missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Trifluoromethoxy)benzyl Bromide 98.0+%, TCI America™
$67.38 - $177.11
Chemical Identifiers
| CAS | 50824-05-0 |
|---|---|
| Molecular Formula | C8H6BrF3O |
| Molecular Weight (g/mol) | 255.034 |
| MDL Number | MFCD00061238 |
| InChI Key | JDNPUJCKXLOHOW-UHFFFAOYSA-N |
| Synonym | 4-trifluoromethoxy benzyl bromide, 1-bromomethyl-4-trifluoromethoxy benzene, 4-trifluoromethoxybenzyl bromide, 4-trifluoromethoxy benzylbromide, p-trifluoromethoxy benzyl bromide, alpha-bromo-4-trifluoromethoxy toluene, benzene, 1-bromomethyl-4-trifluoromethoxy, 4-trifluoromethoxy-benzyl bromide, 4-bromomethyl phenoxy trifluoromethane, 1-bromomethyl-4-trifluoromethoxy-benzene |
| PubChem CID | 142785 |
| IUPAC Name | 1-(bromomethyl)-4-(trifluoromethoxy)benzene |
| SMILES | C1=CC(=CC=C1CBr)OC(F)(F)F |
Chemical Identifiers
| 50824-05-0 | |
| 255.034 | |
| JDNPUJCKXLOHOW-UHFFFAOYSA-N | |
| 142785 | |
| C1=CC(=CC=C1CBr)OC(F)(F)F |
| C8H6BrF3O | |
| MFCD00061238 | |
| 4-trifluoromethoxy benzyl bromide, 1-bromomethyl-4-trifluoromethoxy benzene, 4-trifluoromethoxybenzyl bromide, 4-trifluoromethoxy benzylbromide, p-trifluoromethoxy benzyl bromide, alpha-bromo-4-trifluoromethoxy toluene, benzene, 1-bromomethyl-4-trifluoromethoxy, 4-trifluoromethoxy-benzyl bromide, 4-bromomethyl phenoxy trifluoromethane, 1-bromomethyl-4-trifluoromethoxy-benzene | |
| 1-(bromomethyl)-4-(trifluoromethoxy)benzene |
Specifications
| 50824-05-0 | |
| 79°C | |
| MFCD00061238 | |
| 3261 | |
| JDNPUJCKXLOHOW-UHFFFAOYSA-N | |
| 1-(bromomethyl)-4-(trifluoromethoxy)benzene | |
| 142785 | |
| ≥98.0% (GC) | |
| 4-(Trifluoromethoxy)benzyl Bromide |
| White-Yellow | |
| C8H6BrF3O | |
| 1 g | |
| 4-trifluoromethoxy benzyl bromide, 1-bromomethyl-4-trifluoromethoxy benzene, 4-trifluoromethoxybenzyl bromide, 4-trifluoromethoxy benzylbromide, p-trifluoromethoxy benzyl bromide, alpha-bromo-4-trifluoromethoxy toluene, benzene, 1-bromomethyl-4-trifluoromethoxy, 4-trifluoromethoxy-benzyl bromide, 4-bromomethyl phenoxy trifluoromethane, 1-bromomethyl-4-trifluoromethoxy-benzene | |
| C1=CC(=CC=C1CBr)OC(F)(F)F | |
| 255.034 | |
| 255.03 | |
| Crystalline Lumps |
Safety and Handling
TSCA : No