missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)benzyl bromide, 97%

Catalog No. p-7024120
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1833506 5 g
AAA1833514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1833506 Supplier Thermo Scientific Chemicals Supplier No. A1833506

CAS: 50824-05-0 | C8H6BrF3O | 255.034 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50824-05-0
Molecular Formula C8H6BrF3O
Molecular Weight (g/mol) 255.034
MDL Number MFCD00061238
InChI Key JDNPUJCKXLOHOW-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy benzyl bromide, 1-bromomethyl-4-trifluoromethoxy benzene, 4-trifluoromethoxybenzyl bromide, 4-trifluoromethoxy benzylbromide, p-trifluoromethoxy benzyl bromide, alpha-bromo-4-trifluoromethoxy toluene, benzene, 1-bromomethyl-4-trifluoromethoxy, 4-trifluoromethoxy-benzyl bromide, 4-bromomethyl phenoxy trifluoromethane, 1-bromomethyl-4-trifluoromethoxy-benzene
PubChem CID 142785
IUPAC Name 1-(bromomethyl)-4-(trifluoromethoxy)benzene
SMILES C1=CC(=CC=C1CBr)OC(F)(F)F

Specifications

Melting Point 22°C to 24°C
Density 1.594
Boiling Point 63°C (3 mmHg)
Flash Point 94°C (201°F)
Refractive Index 1.48
Quantity 5 g
UN Number UN3261
Beilstein 2521451
Formula Weight 255.04
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethoxy)benzyl bromide
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only