missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Trifluoromethoxy)benzonitrile 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 193°C |
| Quantity | 1 g |
| UN Number | 3276 |
| Formula Weight | 187.12 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 4-(Trifluoromethoxy)benzonitrile |
Safety and Handling
| TSCA | No |