missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)benzoic Acid 97.0+%, TCI America™

Catalog No. p-7136757
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14001G 1 g
T14005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T14001G Supplier TCI America Supplier No. T14001G

Chemical Identifiers

CAS 330-12-1
Molecular Formula C8H5F3O3
Molecular Weight (g/mol) 206.12
MDL Number MFCD00002541
InChI Key RATSANVPHHXDCT-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy benzoic acid, p-trifluoromethoxybenzoic acid, 4-trifluoromethoxy-benzoic acid, p-carboxyphenyl trifluoromethyl ether, 4-trifluoromethoxy benzoicacid, benzoic acid, 4-trifluoromethoxy, alpha,alpha,alpha-trifluoro-p-anisic acid, pubchem4593, acmc-1cp95, ksc225q9l
PubChem CID 67613
IUPAC Name 4-(trifluoromethoxy)benzoic acid
SMILES C1=CC(=CC=C1C(=O)O)OC(F)(F)F

Specifications

Melting Point 154°C
Color White-Yellow
Quantity 1 g
Formula Weight 206.12
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Trifluoromethoxy)benzoic Acid