missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)benzenesulfonyl chloride, 98%

Catalog No. p-7022608
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1546703 1 g
AAA1546706 5 g
AAA1546714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1546703 Supplier Thermo Scientific Chemicals Supplier No. A1546703

CAS: 94108-56-2 | C7H4ClF3O3S | 260.611 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 94108-56-2
Molecular Formula C7H4ClF3O3S
Molecular Weight (g/mol) 260.611
MDL Number MFCD00042408
InChI Key UHCDBMIOLNKDHG-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy benzenesulfonyl chloride, 4-trifluoromethoxy benzene-1-sulfonyl chloride, 4-trifluoromethoxy benzenesulphonyl chloride, benzenesulfonyl chloride, 4-trifluoromethoxy, p-trifluoromethoxy benzenesulphonyl chloride, 4-trifluoromethoxyphenyl-sulfonylchloride, 4-trifluoromethoxyphenylsulfonyl chloride, 4-trifluoromethoxy benzensulfonylchloride, 4-trifluoromethoxy-benzenesulfonylchloride, 4-trifluoromethoxy-benzenesulfonyl chloride
PubChem CID 523102
IUPAC Name 4-(trifluoromethoxy)benzenesulfonyl chloride
SMILES C1=CC(=CC=C1OC(F)(F)F)S(=O)(=O)Cl

Specifications

Density 1.536
Boiling Point 120°C to 122°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.48
Quantity 1 g
UN Number UN3265
Beilstein 7641524
Sensitivity Moisture sensitive
Formula Weight 260.62
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethoxy)benzenesulfonyl chloride
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 302-397-9
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only