missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)benzaldehyde, 96%

CAS: 659-28-9 | C8H5F3O2 | 190.121 g/mol

$88.10 - $294.81

Chemical Identifiers

CAS 659-28-9
Molecular Formula C8H5F3O2
Molecular Weight (g/mol) 190.121
MDL Number MFCD00041530
InChI Key XQNVDQZWOBPLQZ-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy benzaldehyde, p-trifluoromethoxybenzaldehyde, p-trifluoromethoxy benzaldehyde, benzaldehyde, 4-trifluoromethoxy, 4-trifluoromethoxy-benzaldehyde, alpha,alpha,alpha-trifluoroanisaldehyde, pubchem1471, acmc-209nsx
PubChem CID 69573
IUPAC Name 4-(trifluoromethoxy)benzaldehyde
SMILES C1=CC(=CC=C1C=O)OC(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1067106
View Documents
Thermo Scientific Chemicals
A1067106
5 g
Each for $88.10
Only null left
 
AAA1067114
View Documents
Thermo Scientific Chemicals
A1067114
25 g
Each for $294.81
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

659-28-9
190.121
XQNVDQZWOBPLQZ-UHFFFAOYSA-N
69573
C1=CC(=CC=C1C=O)OC(F)(F)F
C8H5F3O2
MFCD00041530
4-trifluoromethoxy benzaldehyde, p-trifluoromethoxybenzaldehyde, p-trifluoromethoxy benzaldehyde, benzaldehyde, 4-trifluoromethoxy, 4-trifluoromethoxy-benzaldehyde, alpha,alpha,alpha-trifluoroanisaldehyde, pubchem1471, acmc-209nsx
4-(trifluoromethoxy)benzaldehyde

Specifications

659-28-9
93°C (27 mmHg)
C8H5F3O2
MFCD00041530
1949135
4-trifluoromethoxy benzaldehyde, p-trifluoromethoxybenzaldehyde, p-trifluoromethoxy benzaldehyde, benzaldehyde, 4-trifluoromethoxy, 4-trifluoromethoxy-benzaldehyde, alpha,alpha,alpha-trifluoroanisaldehyde, pubchem1471, acmc-209nsx
C1=CC(=CC=C1C=O)OC(F)(F)F
190.121
190.12
4- (Trifluoromethoxy)benzaldehyde
1.331
71°C (159°F)
1.458
5 g
Air Sensitive
XQNVDQZWOBPLQZ-UHFFFAOYSA-N
4-(trifluoromethoxy)benzaldehyde
69573
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P370+P378q-P501c

H227-H302-H315-H319

EINECSNumber : 211-531-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only