missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)benzaldehyde, 96%

Catalog No. AAA1067106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1067106 5 g
AAA1067114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1067106 Supplier Thermo Scientific Chemicals Supplier No. A1067106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 659-28-9 | C8H5F3O2 | 190.121 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 659-28-9
Molecular Formula C8H5F3O2
Molecular Weight (g/mol) 190.121
MDL Number MFCD00041530
InChI Key XQNVDQZWOBPLQZ-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy benzaldehyde, p-trifluoromethoxybenzaldehyde, p-trifluoromethoxy benzaldehyde, benzaldehyde, 4-trifluoromethoxy, 4-trifluoromethoxy-benzaldehyde, alpha,alpha,alpha-trifluoroanisaldehyde, pubchem1471, acmc-209nsx
PubChem CID 69573
IUPAC Name 4-(trifluoromethoxy)benzaldehyde
SMILES C1=CC(=CC=C1C=O)OC(F)(F)F

Specifications

Density 1.331
Boiling Point 93°C (27 mmHg)
Flash Point 71°C (159°F)
Refractive Index 1.458
Quantity 5 g
Beilstein 1949135
Sensitivity Air Sensitive
Formula Weight 190.12
Percent Purity 96%
Chemical Name or Material 4- (Trifluoromethoxy)benzaldehyde
Hazard Category H227-H302-H315-H319
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 211-531-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only