missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)anisole, 98+%

Catalog No. AAA1974614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1974614 25 g
AAA1974606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1974614 Supplier Thermo Scientific Chemicals Supplier No. A1974614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 710-18-9 | C8H7F3O2 | 192.137 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 710-18-9
Molecular Formula C8H7F3O2
Molecular Weight (g/mol) 192.137
MDL Number MFCD00216942
InChI Key NOAFZIOGGDPYKK-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy anisole, 1-methoxy-4-trifluoromethoxy benzene, p-trifluoromethoxy anisole, 4-trifluoromethoxyanisole, benzene, 1-methoxy-4-trifluoromethoxy, 4-methoxy trifluoromethoxy benzene, hydroquinone methyl trifluoromethyl ether, trifluoro 4-methoxyphenoxy methane, pubchem3570, acmc-1bcz3
PubChem CID 2724973
IUPAC Name 1-methoxy-4-(trifluoromethoxy)benzene
SMILES COC1=CC=C(C=C1)OC(F)(F)F

Specifications

Density 1.272
Boiling Point 163°C to 165°C
Flash Point 58°C (136°F)
Refractive Index 1.432
Quantity 25 g
UN Number UN1993
Beilstein 2556809
Formula Weight 192.14
Percent Purity ≥98%
Chemical Name or Material 4-(Trifluoromethoxy)anisole
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only