missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)aniline, 99%

Catalog No. AC297640025
Change view
Click to view available options
Quantity:
2.5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC297640025 2.5 g Glass bottle
AC297640250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC297640025 Supplier Thermo Scientific Chemicals Supplier No. 297640025
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 461-82-5 | C7H6F3NO | 177.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 461-82-5
Molecular Formula C7H6F3NO
Molecular Weight (g/mol) 177.12
MDL Number MFCD00041314
InChI Key XUJFOSLZQITUOI-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy aniline, 4-trifluoromethoxy-phenylamine, 4-trifluoromethoxy benzenamine, p-trifluoromethoxyaniline, 4-trifluoroethoxy aniline, benzenamine, 4-trifluoromethoxy, unii-p40l42cvf6, alpha,alpha,alpha-trifluoro-p-anisidine, p-aminotrifluoromethoxybenzene, 4-amino-alpha,alpha,alpha-trifluoroanisole
PubChem CID 600848
IUPAC Name 4-(trifluoromethoxy)aniline
SMILES C1=CC(=CC=C1N)OC(F)(F)F

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.3100g/mL
Boiling Point 83.0°C to 85.0°C (15.0 mmHg)
Flash Point 80°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4620 to 1.4640
Linear Formula CF3OC6H4NH2
Quantity 2.5 g
Specific Gravity 1.31
Formula Weight 177.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethoxy)aniline, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Toxic if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Fatal in contact with skin.
May cause damage to organs through prolonged or repeated exposure.<b
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
EINECSNumber 207-317-5

RUO – Research Use Only