missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)aniline, 98%

CAS: 461-82-5 | C7H6F3NO | 177.126 g/mol

$60.90 - $210.07

Chemical Identifiers

CAS 461-82-5
Molecular Formula C7H6F3NO
Molecular Weight (g/mol) 177.126
MDL Number MFCD00041314
InChI Key XUJFOSLZQITUOI-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy aniline, 4-trifluoromethoxy-phenylamine, 4-trifluoromethoxy benzenamine, p-trifluoromethoxyaniline, 4-trifluoroethoxy aniline, benzenamine, 4-trifluoromethoxy, unii-p40l42cvf6, alpha,alpha,alpha-trifluoro-p-anisidine, p-aminotrifluoromethoxybenzene, 4-amino-alpha,alpha,alpha-trifluoroanisole
PubChem CID 600848
IUPAC Name 4-(trifluoromethoxy)aniline
SMILES C1=CC(=CC=C1N)OC(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1720806
View Documents
Thermo Scientific Chemicals
A1720806
5 g
Each for $60.90
Only null left
 
AAA1720814
View Documents
Thermo Scientific Chemicals
A1720814
25 g
Each for $210.07
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

461-82-5
177.126
XUJFOSLZQITUOI-UHFFFAOYSA-N
600848
C1=CC(=CC=C1N)OC(F)(F)F
C7H6F3NO
MFCD00041314
4-trifluoromethoxy aniline, 4-trifluoromethoxy-phenylamine, 4-trifluoromethoxy benzenamine, p-trifluoromethoxyaniline, 4-trifluoroethoxy aniline, benzenamine, 4-trifluoromethoxy, unii-p40l42cvf6, alpha,alpha,alpha-trifluoro-p-anisidine, p-aminotrifluoromethoxybenzene, 4-amino-alpha,alpha,alpha-trifluoroanisole
4-(trifluoromethoxy)aniline

Specifications

461-82-5
73°C to 75°C (10 mmHg)
Characteristic
1.463
5 g
2090209
XUJFOSLZQITUOI-UHFFFAOYSA-N
4-(trifluoromethoxy)aniline
600848
98%
1.31
81°C (178°F)
C7H6F3NO
MFCD00041314
UN2810
4-trifluoromethoxy aniline, 4-trifluoromethoxy-phenylamine, 4-trifluoromethoxy benzenamine, p-trifluoromethoxyaniline, 4-trifluoroethoxy aniline, benzenamine, 4-trifluoromethoxy, unii-p40l42cvf6, alpha,alpha,alpha-trifluoro-p-anisidine, p-aminotrifluoromethoxybenzene, 4-amino-alpha,alpha,alpha-trifluoroanisole
C1=CC(=CC=C1N)OC(F)(F)F
177.126
177.13
4-(Trifluoromethoxy)aniline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H310-H318-H373-H227-H315
Toxic if swallowed.
Fatal in contact with skin.
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Causes skin irritation.

P210-P235-P260-P262-P264b-P270-P271-P280-P301+P310-P302+P350-P304+P340-P305+P351+P338-P310-P312-P314-P330-P332+P313-P361-P363-P370+P378q-P501c

H227-H301-H310-H315-H318-H335-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

EINECSNumber : 207-317-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only