missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)aniline, 98%

Catalog No. AAA1720814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1720814 25 g
AAA1720806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1720814 Supplier Thermo Scientific Chemicals Supplier No. A1720814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 461-82-5 | C7H6F3NO | 177.126 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 461-82-5
Molecular Formula C7H6F3NO
Molecular Weight (g/mol) 177.126
MDL Number MFCD00041314
InChI Key XUJFOSLZQITUOI-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy aniline, 4-trifluoromethoxy-phenylamine, 4-trifluoromethoxy benzenamine, p-trifluoromethoxyaniline, 4-trifluoroethoxy aniline, benzenamine, 4-trifluoromethoxy, unii-p40l42cvf6, alpha,alpha,alpha-trifluoro-p-anisidine, p-aminotrifluoromethoxybenzene, 4-amino-alpha,alpha,alpha-trifluoroanisole
PubChem CID 600848
IUPAC Name 4-(trifluoromethoxy)aniline
SMILES C1=CC(=CC=C1N)OC(F)(F)F

Specifications

Density 1.31
Boiling Point 73°C to 75°C (10 mmHg)
Flash Point 81°C (178°F)
Odor Characteristic
Refractive Index 1.463
Quantity 25 g
UN Number UN2810
Beilstein 2090209
Formula Weight 177.13
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethoxy)aniline
Show More Show Less
Hazard Category H227-H301-H310-H315-H318-H335-H373
Hazard Statement GHS H Statement
H301-H310-H318-H373-H227-H315
Toxic if swallowed.
Fatal in contact with skin.
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Causes skin irritation.
Precautionary Statement P210-P235-P260-P262-P264b-P270-P271-P280-P301+P310-P302+P350-P304+P340-P305+P351+P338-P310-P312-P314-P330-P332+P313-P361-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 207-317-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only