missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Trifluoromethoxy)aniline 98.0+%, TCI America™
$44.28 - $90.48
Chemical Identifiers
| CAS | 461-82-5 |
|---|---|
| Molecular Formula | C7H6F3NO |
| Molecular Weight (g/mol) | 177.126 |
| MDL Number | MFCD00041314 |
| InChI Key | XUJFOSLZQITUOI-UHFFFAOYSA-N |
| Synonym | 4-trifluoromethoxy aniline, 4-trifluoromethoxy-phenylamine, 4-trifluoromethoxy benzenamine, p-trifluoromethoxyaniline, 4-trifluoroethoxy aniline, benzenamine, 4-trifluoromethoxy, unii-p40l42cvf6, alpha,alpha,alpha-trifluoro-p-anisidine, p-aminotrifluoromethoxybenzene, 4-amino-alpha,alpha,alpha-trifluoroanisole |
| PubChem CID | 600848 |
| IUPAC Name | 4-(trifluoromethoxy)aniline |
| SMILES | C1=CC(=CC=C1N)OC(F)(F)F |
Chemical Identifiers
| 461-82-5 | |
| 177.126 | |
| XUJFOSLZQITUOI-UHFFFAOYSA-N | |
| 600848 | |
| C1=CC(=CC=C1N)OC(F)(F)F |
| C7H6F3NO | |
| MFCD00041314 | |
| 4-trifluoromethoxy aniline, 4-trifluoromethoxy-phenylamine, 4-trifluoromethoxy benzenamine, p-trifluoromethoxyaniline, 4-trifluoroethoxy aniline, benzenamine, 4-trifluoromethoxy, unii-p40l42cvf6, alpha,alpha,alpha-trifluoro-p-anisidine, p-aminotrifluoromethoxybenzene, 4-amino-alpha,alpha,alpha-trifluoroanisole | |
| 4-(trifluoromethoxy)aniline |
Specifications
| 461-82-5 | |
| C7H6F3NO | |
| 5 g | |
| 4-trifluoromethoxy aniline, 4-trifluoromethoxy-phenylamine, 4-trifluoromethoxy benzenamine, p-trifluoromethoxyaniline, 4-trifluoroethoxy aniline, benzenamine, 4-trifluoromethoxy, unii-p40l42cvf6, alpha,alpha,alpha-trifluoro-p-anisidine, p-aminotrifluoromethoxybenzene, 4-amino-alpha,alpha,alpha-trifluoroanisole | |
| C1=CC(=CC=C1N)OC(F)(F)F | |
| 177.126 | |
| 177.13 | |
| Liquid |
| 83°C | |
| MFCD00041314 | |
| 2810 | |
| XUJFOSLZQITUOI-UHFFFAOYSA-N | |
| 4-(trifluoromethoxy)aniline | |
| 600848 | |
| ≥98.0% (GC,T) | |
| 4-(Trifluoromethoxy)aniline |
Safety and Handling
TSCA : No