Learn More
4-(Tri-n-butylstannyl)thiazole, 95%
CAS: 173979-01-6 | C15H29NSSn | 374.17 g/mol
Supplier: Thermo Scientific Chemicals H5145703
Description
4-(Tri-n-butylstannyl)thiazole is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-(Tri-n-butylstannyl)thiazole is used as pharmaceutical intermediate.
Solubility
Not miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
| 4-(Tri-n-butylstannyl)thiazole | |
| C15H29NSSn | |
| 1 g | |
| 4-tributylstannyl thiazole, 4-tributylstannyl-1,3-thiazole, 4-tributylstannylthiazole, 4-tri-n-butylstannyl thiazole, 4-tributylstannanyl-1,3-thiazole, 4-tributylstannanyl-thiazole, 4-tri-n-butylstannylthiazole, 4-thiazolyltri-n-butylstannane, tri-n-butyl-4-thiazolyltin | |
| YYQKQPYPLADFMK-UHFFFAOYSA-N | |
| 4-(tributylstannyl)-1,3-thiazole | |
| 2763258 | |
| 95% |
| 173979-01-6 | |
| MFCD01319073 | |
| UN2788 | |
| Not miscible in water. | |
| CCCC[Sn](CCCC)(CCCC)C1=CSC=N1 | |
| 374.17 | |
| 374.17 |
Chemical Identifiers
| 173979-01-6 | |
| 374.17 | |
| YYQKQPYPLADFMK-UHFFFAOYSA-N | |
| 2763258 |
| C15H29NSSn | |
| MFCD01319073 | |
| 4-tributylstannyl thiazole, 4-tributylstannyl-1,3-thiazole, 4-tributylstannylthiazole, 4-tri-n-butylstannyl thiazole, 4-tributylstannanyl-1,3-thiazole, 4-tributylstannanyl-thiazole, 4-tri-n-butylstannylthiazole, 4-thiazolyltri-n-butylstannane, tri-n-butyl-4-thiazolyltin | |
| CCCC[Sn](CCCC)(CCCC)C1=CSC=N1 |
Safety and Handling
GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
H301-H312-H315-H319-H372
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only