missing translation for 'onlineSavingsMsg'
Learn More

4-(Tri-n-butylstannyl)thiazole, 95%

Catalog No. p-7033992
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH51457MD 250 mg
AAH5145703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAH51457MD Supplier Thermo Scientific Chemicals Supplier No. H51457MD
Only null left
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 173979-01-6 | C15H29NSSn | 374.17 g/mol

4-(Tri-n-butylstannyl)thiazole is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Tri-n-butylstannyl)thiazole is used as pharmaceutical intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 173979-01-6
Molecular Formula C15H29NSSn
Molecular Weight (g/mol) 374.17
MDL Number MFCD01319073
InChI Key YYQKQPYPLADFMK-UHFFFAOYSA-N
Synonym 4-tributylstannyl thiazole, 4-tributylstannyl-1,3-thiazole, 4-tributylstannylthiazole, 4-tri-n-butylstannyl thiazole, 4-tributylstannanyl-1,3-thiazole, 4-tributylstannanyl-thiazole, 4-tri-n-butylstannylthiazole, 4-thiazolyltri-n-butylstannane, tri-n-butyl-4-thiazolyltin
PubChem CID 2763258
SMILES CCCC[Sn](CCCC)(CCCC)C1=CSC=N1

Specifications

Quantity 250 mg
UN Number UN2788
Solubility Information Not miscible in water.
IUPAC Name tributyl(1,3-thiazol-4-yl)stannane
Formula Weight 374.17
Percent Purity 95%
Chemical Name or Material 4-(Tri-n-butylstannyl)thiazole
Hazard Category H301-H312-H315-H319-H372
Hazard Statement GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only