missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(trans-4-Propylcyclohexyl)cyclohexanone 98.0+%, TCI America™
Supplier: TCI America P21085G
Specifications
| 4-(trans-4-Propylcyclohexyl)cyclohexanone | |
| White | |
| MFCD04038797 | |
| 4-trans-4-propylcyclohexyl cyclohexanone, 4-4-propylcyclohexyl cyclohexanone, 4-propyldicyclohexylanone, 4-trans-4'-n-propylcyclohexyl-cyclohexanone, 4-4-propylcyclohexyl cyclohexan-1-one, 4-trans-4'-propylcyclohexyl cyclohexanone, 4'-propyl-1,1'-bicyclohexyl-4-one, 4'-propyl bi cyclohexyl-4-one, 1's,4'r-4'-propyl-1,1'-bi cyclohexane-4-one, pubchem2509 | |
| CCCC1CCC(CC1)C2CCC(=O)CC2 | |
| 222.372 | |
| 222.37 | |
| Crystalline Powder |
| 82832-73-3 | |
| C15H26O | |
| 5 g | |
| AKCZQKBKWXBJOF-UHFFFAOYSA-N | |
| 4-(4-propylcyclohexyl)cyclohexan-1-one | |
| 4354185 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 82832-73-3 | |
| 222.372 | |
| AKCZQKBKWXBJOF-UHFFFAOYSA-N | |
| 4354185 | |
| CCCC1CCC(CC1)C2CCC(=O)CC2 |
| C15H26O | |
| MFCD04038797 | |
| 4-trans-4-propylcyclohexyl cyclohexanone, 4-4-propylcyclohexyl cyclohexanone, 4-propyldicyclohexylanone, 4-trans-4'-n-propylcyclohexyl-cyclohexanone, 4-4-propylcyclohexyl cyclohexan-1-one, 4-trans-4'-propylcyclohexyl cyclohexanone, 4'-propyl-1,1'-bicyclohexyl-4-one, 4'-propyl bi cyclohexyl-4-one, 1's,4'r-4'-propyl-1,1'-bi cyclohexane-4-one, pubchem2509 | |
| 4-(4-propylcyclohexyl)cyclohexan-1-one |
Safety and Handling
EINECSNumber : (4)-1698
TSCA : No