missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(trans-4-Propylcyclohexyl)cyclohexanone 98.0+%, TCI America™
$48.13 - $123.21
Chemical Identifiers
| CAS | 82832-73-3 |
|---|---|
| Molecular Formula | C15H26O |
| Molecular Weight (g/mol) | 222.372 |
| MDL Number | MFCD04038797 |
| InChI Key | AKCZQKBKWXBJOF-UHFFFAOYSA-N |
| Synonym | 4-trans-4-propylcyclohexyl cyclohexanone, 4-4-propylcyclohexyl cyclohexanone, 4-propyldicyclohexylanone, 4-trans-4'-n-propylcyclohexyl-cyclohexanone, 4-4-propylcyclohexyl cyclohexan-1-one, 4-trans-4'-propylcyclohexyl cyclohexanone, 4'-propyl-1,1'-bicyclohexyl-4-one, 4'-propyl bi cyclohexyl-4-one, 1's,4'r-4'-propyl-1,1'-bi cyclohexane-4-one, pubchem2509 |
| PubChem CID | 4354185 |
| IUPAC Name | 4-(4-propylcyclohexyl)cyclohexan-1-one |
| SMILES | CCCC1CCC(CC1)C2CCC(=O)CC2 |
Chemical Identifiers
| 82832-73-3 | |
| 222.372 | |
| AKCZQKBKWXBJOF-UHFFFAOYSA-N | |
| 4354185 | |
| CCCC1CCC(CC1)C2CCC(=O)CC2 |
| C15H26O | |
| MFCD04038797 | |
| 4-trans-4-propylcyclohexyl cyclohexanone, 4-4-propylcyclohexyl cyclohexanone, 4-propyldicyclohexylanone, 4-trans-4'-n-propylcyclohexyl-cyclohexanone, 4-4-propylcyclohexyl cyclohexan-1-one, 4-trans-4'-propylcyclohexyl cyclohexanone, 4'-propyl-1,1'-bicyclohexyl-4-one, 4'-propyl bi cyclohexyl-4-one, 1's,4'r-4'-propyl-1,1'-bi cyclohexane-4-one, pubchem2509 | |
| 4-(4-propylcyclohexyl)cyclohexan-1-one |
Specifications
| 82832-73-3 | |
| C15H26O | |
| 1 g | |
| AKCZQKBKWXBJOF-UHFFFAOYSA-N | |
| 4-(4-propylcyclohexyl)cyclohexan-1-one | |
| 4354185 | |
| ≥98.0% (GC) | |
| 4-(trans-4-Propylcyclohexyl)cyclohexanone |
| White | |
| MFCD04038797 | |
| 4-trans-4-propylcyclohexyl cyclohexanone, 4-4-propylcyclohexyl cyclohexanone, 4-propyldicyclohexylanone, 4-trans-4'-n-propylcyclohexyl-cyclohexanone, 4-4-propylcyclohexyl cyclohexan-1-one, 4-trans-4'-propylcyclohexyl cyclohexanone, 4'-propyl-1,1'-bicyclohexyl-4-one, 4'-propyl bi cyclohexyl-4-one, 1's,4'r-4'-propyl-1,1'-bi cyclohexane-4-one, pubchem2509 | |
| CCCC1CCC(CC1)C2CCC(=O)CC2 | |
| 222.372 | |
| 222.37 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (4)-1698
TSCA : No