missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(trans-4-Pentylcyclohexyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$193.86 - $193.86
Chemical Identifiers
| CAS | 143651-26-7 |
|---|---|
| Molecular Formula | C17H27BO2 |
| MDL Number | MFCD09750938 |
| InChI Key | JFESOTHKCUMHGA-UHFFFAOYSA-N |
| Synonym | 4-(trans-4-Pentylcyclohexyl)benzeneboronic Acid |
| PubChem CID | 3696091 |
| IUPAC Name | [4-(4-pentylcyclohexyl)phenyl]boronic acid |
Chemical Identifiers
| 143651-26-7 | |
| MFCD09750938 | |
| 4-(trans-4-Pentylcyclohexyl)benzeneboronic Acid | |
| [4-(4-pentylcyclohexyl)phenyl]boronic acid |
| C17H27BO2 | |
| JFESOTHKCUMHGA-UHFFFAOYSA-N | |
| 3696091 |
Specifications
| 143651-26-7 | |
| C17H27BO2 | |
| 5 g | |
| JFESOTHKCUMHGA-UHFFFAOYSA-N | |
| [4-(4-pentylcyclohexyl)phenyl]boronic acid | |
| 3696091 | |
| Crystalline Powder |
| White | |
| MFCD09750938 | |
| 4-(trans-4-Pentylcyclohexyl)benzeneboronic Acid | |
| B(C1=CC=C(C=C1)C2CCC(CC2)CCCCC)(O)O | |
| 274.211 | |
| 274.21 | |
| 4-(trans-4-Pentylcyclohexyl)phenylboronic Acid (contains varying amounts of Anhydride) |
Safety and Handling
TSCA : No