accessibility menu, dialog, popup

UserName

BCH3OHHO

CAS RN 143651-26-7

IUPAC Name: [4-(4-pentylcyclohexyl)phenyl]boronic acid
Synonyms: 4-(4-pentylcyclohexyl)phenylboronic acid
Molecular Weight (g/mol): 274.21
Molecular Formula: C17H27BO2
InChi Key: JFESOTHKCUMHGA-UHFFFAOYSA-N
SMILES: CCCCCC1CCC(CC1)C1=CC=C(C=C1)B(O)O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results