missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(trans-4-Pentylcyclohexyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 5 g |
| SMILES | B(C1=CC=C(C=C1)C2CCC(CC2)CCCCC)(O)O |
| Molecular Weight (g/mol) | 274.211 |
| Formula Weight | 274.21 |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-(trans-4-Pentylcyclohexyl)phenylboronic Acid (contains varying amounts of Anhydride) |
Safety and Handling
| TSCA | No |