missing translation for 'onlineSavingsMsg'
Learn More

4'-(trans-4-Amylcyclohexyl)biphenyl-4-carbonitrile 98.0+%, TCI America™

Catalog No. p-7117974
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25681G 1 g
A25685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A25681G Supplier TCI America Supplier No. A25681G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 68065-81-6
Molecular Formula C24H29N
Molecular Weight (g/mol) 331.503
MDL Number MFCD01076317
InChI Key QKEBUASRTJNJJS-UHFFFAOYSA-N
Synonym 4'-trans-4-pentylcyclohexyl-1,1'-biphenyl-4-carbonitrile, trans-4'-4-pentylcyclohexyl-4-biphenylcarbonitrile, trans-4-4-pentylcyclohexyl-4'-cyanobiphenyl, 4-trans-4-n-pentylcyclohexyl biphenyl-4'-carbonitrile, 4-4-4-pentylcyclohexyl phenyl benzonitrile, 1,1'-biphenyl-4-carbonitrile, 4'-trans-4-pentylcyclohexyl, 4-cyano-4'-trans-4-pentylcyclohexyl biphenyl, 4'-trans-4-amylcyclohexyl biphenyl-4-carbonitrile, trans-4'-4-pentylcyclohexyl 1,1'-biphenyl-4-carbonitrile
PubChem CID 106491
IUPAC Name 4-[4-(4-pentylcyclohexyl)phenyl]benzonitrile
SMILES CCCCCC1CCC(CC1)C2=CC=C(C=C2)C3=CC=C(C=C3)C#N

Specifications

Melting Point 96°C
Color White
Boiling Point 211°C
Quantity 1 g
Formula Weight 331.50
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-(trans-4-Amylcyclohexyl)biphenyl-4-carbonitrile