missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Thien-2-yltetrahydropyran-4-carboxylic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC62201DA
Specifications
| 4-thien-2-yltetrahydropyran-4-carboxylic acid | |
| 97% | |
| C10H12O3S | |
| 1g | |
| VWYOZQWRYINSBP-UHFFFAOYSA-N | |
| 4-(thiophen-2-yl)oxane-4-carboxylic acid | |
| 19066254 | |
| 97% |
| 880166-18-7 | |
| Amber Glass Bottle | |
| MFCD09817516 | |
| 4-thiophen-2-yl oxane-4-carboxylic acid, 4-thien-2-yltetrahydropyran-4-carboxylic acid, 4-thien-2-yltetrahydro-2h-pyran-4-carboxylic acid, 2h-pyran-4-carboxylicacid, tetrahydro-4-2-thienyl, 4-thiophen-2-yl tetrahydro-2h-pyran-4-carboxylic acid, 4-thiophen-2-yl oxan-4-carboxylic acid, 4-2-thienyl tetrahydropyran-4-carboxylic acid, 4-thien-2-yl tetrahydro-2h-pyran-4-carboxylic acid, 4-2-thienyl-2h-3,4,5,6-tetrahydropyran-4-carboxylic acid | |
| OC(=O)C1(CCOCC1)C1=CC=CS1 | |
| 212.26 | |
| 212.27 |
Chemical Identifiers
| 880166-18-7 | |
| 212.26 | |
| VWYOZQWRYINSBP-UHFFFAOYSA-N | |
| 19066254 | |
| OC(=O)C1(CCOCC1)C1=CC=CS1 |
| C10H12O3S | |
| MFCD09817516 | |
| 4-thiophen-2-yl oxane-4-carboxylic acid, 4-thien-2-yltetrahydropyran-4-carboxylic acid, 4-thien-2-yltetrahydro-2h-pyran-4-carboxylic acid, 2h-pyran-4-carboxylicacid, tetrahydro-4-2-thienyl, 4-thiophen-2-yl tetrahydro-2h-pyran-4-carboxylic acid, 4-thiophen-2-yl oxan-4-carboxylic acid, 4-2-thienyl tetrahydropyran-4-carboxylic acid, 4-thien-2-yl tetrahydro-2h-pyran-4-carboxylic acid, 4-2-thienyl-2h-3,4,5,6-tetrahydropyran-4-carboxylic acid | |
| 4-(thiophen-2-yl)oxane-4-carboxylic acid |
Safety and Handling
GHS Signal Word: Warning