missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Thien-2-yltetrahydropyran-4-carboxylic acid, 97%, Thermo Scientific™
Specifications
Specifications
| Assay Percent Range | 97% |
| Packaging | Amber Glass Bottle |
| Quantity | 250mg |
| Formula Weight | 212.27 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-thien-2-yltetrahydropyran-4-carboxylic acid |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |