missing translation for 'onlineSavingsMsg'
Learn More

4-Thien-2-yltetrahydropyran-4-carboxylic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC62201CB

 View more versions of this product

Catalog No. CC62201CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

4-thien-2-yltetrahydropyran-4-carboxylic acid
97%
C10H12O3S
250mg
VWYOZQWRYINSBP-UHFFFAOYSA-N
4-thiophen-2-yloxane-4-carboxylic acid
19066254
97%
880166-18-7
Amber Glass Bottle
MFCD09817516
4-thiophen-2-yl oxane-4-carboxylic acid, 4-thien-2-yltetrahydropyran-4-carboxylic acid, 4-thien-2-yltetrahydro-2h-pyran-4-carboxylic acid, 2h-pyran-4-carboxylicacid, tetrahydro-4-2-thienyl, 4-thiophen-2-yl tetrahydro-2h-pyran-4-carboxylic acid, 4-thiophen-2-yl oxan-4-carboxylic acid, 4-2-thienyl tetrahydropyran-4-carboxylic acid, 4-thien-2-yl tetrahydro-2h-pyran-4-carboxylic acid, 4-2-thienyl-2h-3,4,5,6-tetrahydropyran-4-carboxylic acid
OC(=O)C1(CCOCC1)C1=CC=CS1
212.26
212.27

Chemical Identifiers

880166-18-7
212.26
VWYOZQWRYINSBP-UHFFFAOYSA-N
19066254
OC(=O)C1(CCOCC1)C1=CC=CS1
C10H12O3S
MFCD09817516
4-thiophen-2-yl oxane-4-carboxylic acid, 4-thien-2-yltetrahydropyran-4-carboxylic acid, 4-thien-2-yltetrahydro-2h-pyran-4-carboxylic acid, 2h-pyran-4-carboxylicacid, tetrahydro-4-2-thienyl, 4-thiophen-2-yl tetrahydro-2h-pyran-4-carboxylic acid, 4-thiophen-2-yl oxan-4-carboxylic acid, 4-2-thienyl tetrahydropyran-4-carboxylic acid, 4-thien-2-yl tetrahydro-2h-pyran-4-carboxylic acid, 4-2-thienyl-2h-3,4,5,6-tetrahydropyran-4-carboxylic acid
4-thiophen-2-yloxane-4-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions