missing translation for 'onlineSavingsMsg'
Learn More

(4-thien-2-ylphenyl)methanol, Thermo Scientific™

Catalog No. p-4391277
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC12009CB 250mg Amber Glass Bottle
CC12009DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC12009CB Supplier Thermo Scientific Chemicals Supplier No. CC12009CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 81443-44-9
Molecular Formula C11H10OS
Molecular Weight (g/mol) 190.26
InChI Key KHZSWSHUIYOMSH-UHFFFAOYSA-N
Synonym 4-thien-2-ylphenyl methanol, 4-thiophen-2-yl phenyl methanol, thiophene,2-4-methylphenyl, benzenemethanol, 4-2-thienyl, 4-thien-2-yl-phenyl methanol, pubchem24308, 4-2-thienyl benzyl alcohol, 4-thien-2-yl benzyl alcohol, 4-2-thienyl phenyl methanol
PubChem CID 2795254
IUPAC Name (4-thiophen-2-ylphenyl)methanol
SMILES C1=CSC(=C1)C2=CC=C(C=C2)CO

Specifications

Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 190.26
Percent Purity 97%
Chemical Name or Material (4-thien-2-ylphenyl)methanol