missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Octylphenol-ring-13C6 Solution, 10 μg/mL in Acetone, MilliporeSigma™ Supelco™

Catalog No. 111018293
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018293 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018293 Supplier Merck Emd Millipore Supplier No. 335651ML

Certan Vial

Environmental

Specifications

CAS 1173020-24-0
Flash Point -17°C - Information refers to the main°Component.
Linear Formula 13C6C8H22 O
MDL Number MFCD09842828
Synonym 4-(1,1,3,3-Tetramethylbutyl)phenol-13C6; 4-tert-OP-13C6
Type Analytical Standard
Quantity 1 mL
Formula Weight 212.28
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 10 μg/mL in acetone