missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-tert-Octylphenol-3,5-d2 Solution, 10 μg/mL in Acetone, MilliporeSigma™ Supelco™
Description
Environmental
Specifications
Specifications
| CAS | 1173021-20-9 |
| Flash Point | -17°C - Information refers to the main°Component. |
| Linear Formula | C14D2H20 O |
| MDL Number | MFCD09842826 |
| Synonym | 4-(1,1,3,3-Tetramethylbutyl)phenol-3,5-d2; 4-tert-OP-D2 |
| Type | Analytical Standard |
| Quantity | 1 mL |
| Formula Weight | 208.34 |
| Grade | Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 10 μg/mL in acetone |