missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Octylphenol-3,5-d2 monoethoxylate Solution, 10 μg/mL in Acetone, MilliporeSigma™ Supelco™

Catalog No. 111018295
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018295 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018295 Supplier Merck Emd Millipore Supplier No. 335231ML
Only null left

Certan Vial

Environmental

Specifications

CAS 1173022-57-5
Linear Formula C16D2H24O2
MDL Number MFCD09842825
Synonym 2-[4-(1,1,3,3-Tetramethylbutyl)phenoxy-3,5-d2 ]ethanol; 4-tert-OP1 EO-D2
Type Analytical Standard
Quantity 1 mL
Formula Weight 252.39
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 10 μg/mL in acetone