missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-tert-Octylphenol-3,5-d2 diethoxylate Solution, 10 μg/mL in Acetone, MilliporeSigma™ Supelco™
Description
Environmental
Specifications
Specifications
| CAS | 1173019-49-2 |
| Flash Point | -17.0°C - Closed Cup |
| Linear Formula | C18D2H28O3 |
| MDL Number | MFCD09953759 |
| Synonym | 2-{2-[4-(1,1,3,3-Tetramethylbutyl)phenoxyl-3,5-d2 ]ethoxy}ethanol; 4-tert-OP2 EO-D2 |
| Type | Analytical Standard |
| Quantity | 1 mL |
| Formula Weight | 296.44 |
| Grade | Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 10 μg/mL in acetone |