missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-tert-Butyltoluene 95.0+%, TCI America™
$41.02 - $87.60
Chemical Identifiers
| CAS | 98-51-1 |
|---|---|
| Molecular Formula | C11H16 |
| Molecular Weight (g/mol) | 148.249 |
| MDL Number | MFCD00008837 |
| InChI Key | QCWXDVFBZVHKLV-UHFFFAOYSA-N |
| Synonym | 4-tert-butyltoluene, p-tert-butyltoluene, 1-tert-butyl-4-methylbenzene, p-t-butyltoluene, benzene, 1-1,1-dimethylethyl-4-methyl, 4-t-butyltoluene, p-methyl-tert-butylbenzene, toluene, p-tert-butyl, 1-methyl-4-tert-butylbenzene, 4-methyl-tert-butylbenzene |
| PubChem CID | 7390 |
| IUPAC Name | 1-tert-butyl-4-methylbenzene |
| SMILES | CC1=CC=C(C=C1)C(C)(C)C |
Chemical Identifiers
| 98-51-1 | |
| 148.249 | |
| QCWXDVFBZVHKLV-UHFFFAOYSA-N | |
| 7390 | |
| CC1=CC=C(C=C1)C(C)(C)C |
| C11H16 | |
| MFCD00008837 | |
| 4-tert-butyltoluene, p-tert-butyltoluene, 1-tert-butyl-4-methylbenzene, p-t-butyltoluene, benzene, 1-1,1-dimethylethyl-4-methyl, 4-t-butyltoluene, p-methyl-tert-butylbenzene, toluene, p-tert-butyl, 1-methyl-4-tert-butylbenzene, 4-methyl-tert-butylbenzene | |
| 1-tert-butyl-4-methylbenzene |
Specifications
| 98-51-1 | |
| 193°C | |
| MFCD00008837 | |
| 2667 | |
| QCWXDVFBZVHKLV-UHFFFAOYSA-N | |
| 1-tert-butyl-4-methylbenzene | |
| 7390 | |
| ≥95.0% (GC) | |
| 4-tert-Butyltoluene |
| Yellow | |
| C11H16 | |
| 25 mL | |
| 4-tert-butyltoluene, p-tert-butyltoluene, 1-tert-butyl-4-methylbenzene, p-t-butyltoluene, benzene, 1-1,1-dimethylethyl-4-methyl, 4-t-butyltoluene, p-methyl-tert-butylbenzene, toluene, p-tert-butyl, 1-methyl-4-tert-butylbenzene, 4-methyl-tert-butylbenzene | |
| CC1=CC=C(C=C1)C(C)(C)C | |
| 148.249 | |
| 148.25 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-0015
RTECSNumber : XS8400000
TSCA : Yes