missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butyltoluene, 96%

Catalog No. AC296320250
Change view
Click to view available options
Quantity:
25 mL
1 L
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC296320250 25 mL Glass bottle
AC296320010 1 L Glass bottle
AC296320025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC296320250 Supplier Thermo Scientific Chemicals Supplier No. 296320250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 98-51-1 | C11H16 | 148.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 98-51-1
Molecular Formula C11H16
Molecular Weight (g/mol) 148.25
MDL Number MFCD00008837
InChI Key QCWXDVFBZVHKLV-UHFFFAOYSA-N
Synonym 4-tert-butyltoluene, p-tert-butyltoluene, 1-tert-butyl-4-methylbenzene, p-t-butyltoluene, benzene, 1-1,1-dimethylethyl-4-methyl, 4-t-butyltoluene, p-methyl-tert-butylbenzene, toluene, p-tert-butyl, 1-methyl-4-tert-butylbenzene, 4-methyl-tert-butylbenzene
PubChem CID 7390
IUPAC Name 1-tert-butyl-4-methylbenzene
SMILES CC1=CC=C(C=C1)C(C)(C)C

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point -54.0°C
Color Colorless to Yellow
Density 0.8612g/mL
Boiling Point 189.0°C to 192.0°C
Flash Point 54°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4910 to 1.4930
Linear Formula (CH3)3CC6H4CH3
Quantity 25 mL
Beilstein 05, 439
Specific Gravity 0.8612
Solubility Information Solubility in water: 0.06 wt% (20°C). Other solubilities: miscible with isopropanol,toluene and cyclohexane,soluble in nearly all organic solvents
Viscosity 1.25 mPa.s (25°C)
Formula Weight 148.25
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 4-tert-Butyltoluene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Fatal if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-675-9

RUO – Research Use Only