missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylstyrene (stabilized with TBC) 90.0+%, TCI America™

Catalog No. B187725ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B187725ML 25 mL
B1877500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B187725ML Supplier TCI America Supplier No. B187725ML

Chemical Identifiers

CAS 1746-23-2
Molecular Formula C12H16
Molecular Weight (g/mol) 160.26
MDL Number MFCD00065126
InChI Key QEDJMOONZLUIMC-UHFFFAOYSA-N
Synonym 4-tert-butylstyrene, p-tert-butylstyrene, p-t-butylstyrene, benzene, 1-1,1-dimethylethyl-4-ethenyl, styrene, p-tert-butyl, unii-9pct9cbw0v, 1-1,1-dimethylethyl-4-ethenylbenzene, 9pct9cbw0v, 1-tert-butyl-4-vinylbenzene, 4-t-butyl styrene
PubChem CID 15627
IUPAC Name 1-tert-butyl-4-ethenylbenzene
SMILES CC(C)(C)C1=CC=C(C=C1)C=C

Specifications

Color Yellow
Quantity 25 mL
Formula Weight 160.26
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-tert-Butylstyrene (stabilized with TBC)
TSCA Yes
Recommended Storage Freezer