missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylpyridine

Catalog No. AC161940250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC161940250 25 g
AC161941000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC161940250 Supplier Thermo Scientific Chemicals Supplier No. 161940250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3978-81-2 | C9H13N | 135.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3978-81-2
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
InChI Key YSHMQTRICHYLGF-UHFFFAOYSA-N
IUPAC Name 4-tert-butylpyridine
SMILES CC(C)(C)C1=CC=NC=C1

Specifications

Density 0.9230 g/mL
Boiling Point 196.0°C to 197.0°C
Flash Point 63°C
Linear Formula (C5H4N)C(CH3)3
Quantity 25 g
Beilstein 20, 252
Specific Gravity 0.923
Solubility Information Solubility aq. soln.: 6.9 g/l (20°C).
Formula Weight 135.22
Chemical Name or Material 4-tert-Butylpyridine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement May cause respiratory irritation.

Causes skin irritation.

Causes serious eye irritation.

Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Wear protective gloves/protective clothing/eye protection/face protection.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician if you feel unwell.

EINECSNumber 223-614-2

RUO – Research Use Only