CAS RN 3978-81-2
IUPAC Name:
4-tert-butylpyridine
Synonyms:
4-(tert-butyl)pyridine
Molecular Weight (g/mol):
135.21
Molecular Formula:
C9H13N
InChi Key:
YSHMQTRICHYLGF-UHFFFAOYSA-N
SMILES:
CC(C)(C)C1=CC=NC=C1
1
–
2
of
2
results
1
4-tert-Butylpyridine
CAS: 3978-81-2 Molecular Formula: C9H13N Molecular Weight (g/mol): 135.21 InChI Key: YSHMQTRICHYLGF-UHFFFAOYSA-N IUPAC Name: 4-tert-butylpyridine SMILES: CC(C)(C)C1=CC=NC=C1
2
4-tert-Butylpyridine 96.0+%, TCI America™
CAS: 3978-81-2 Molecular Formula: C9H13N Molecular Weight (g/mol): 135.21 MDL Number: MFCD00006435 InChI Key: YSHMQTRICHYLGF-UHFFFAOYSA-N Synonym: 4-tert-butyl pyridine,pyridine, 4-1,1-dimethylethyl,4-t-butylpyridine,pyridine, 4-tert-butyl,p-tert-butyl pyridine,unii-il8v02163i,4-tert-butyl-pyridine,4-t-butyl pyridine,4-tert.butylpyridine PubChem CID: 19878 IUPAC Name: 4-tert-butylpyridine SMILES: CC(C)(C)C1=CC=NC=C1