missing translation for 'onlineSavingsMsg'
Learn More

4'-tert-Butylpropiophenone 95.0+%, TCI America™

Catalog No. B16095ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16095ML 5 mL
B16091ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16095ML Supplier TCI America Supplier No. B16095ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 71209-71-7
Molecular Formula C13H18O
Molecular Weight (g/mol) 190.286
MDL Number MFCD00100434
InChI Key AQNYEAINONORRY-UHFFFAOYSA-N
Synonym 4-tert-Butylpropionylbenzene
PubChem CID 595281
IUPAC Name 1-(4-tert-butylphenyl)propan-1-one
SMILES CCC(=O)C1=CC=C(C=C1)C(C)(C)C

Specifications

Color Yellow
Boiling Point 162°C
Quantity 5 mL
UN Number 1993
Formula Weight 190.29
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4′-tert-Butylpropiophenone