accessibility menu, dialog, popup

UserName

CH3OCH3H3CCH3

CAS RN 71209-71-7

IUPAC Name: 1-(4-tert-butylphenyl)propan-1-one
Synonyms: 1-(4-tert-butylphenyl)propan-1-one
Molecular Weight (g/mol): 190.29
Molecular Formula: C13H18O
InChi Key: AQNYEAINONORRY-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC=C(C=C1)C(C)(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results