missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-tert-Butylphthalic Anhydride 98.0+%, TCI America™
$154.02 - $435.08
Chemical Identifiers
| CAS | 32703-79-0 |
|---|---|
| Molecular Formula | C12H12O3 |
| Molecular Weight (g/mol) | 204.23 |
| MDL Number | MFCD00060127 |
| InChI Key | YLJYVKLZVHWUCT-UHFFFAOYSA-N |
| Synonym | 4-tert-butylphthalic anhydride, 5-tert-butyl isobenzofuran-1,3-dione, 1,3-isobenzofurandione, 5-1,1-dimethylethyl, 4-t-butylphthalic anhydride, acmc-1ao20, 4-t-butyl phthalic anhydride, 4-t-butyl-phthalic anhydride, 4-tert-butylphthalicanhydride, 5-tert-butyl-2-benzo b furan-1,3-dione |
| PubChem CID | 122930 |
| IUPAC Name | 5-tert-butyl-1,3-dihydro-2-benzofuran-1,3-dione |
| SMILES | CC(C)(C)C1=CC=C2C(=O)OC(=O)C2=C1 |
Chemical Identifiers
| 32703-79-0 | |
| 204.23 | |
| YLJYVKLZVHWUCT-UHFFFAOYSA-N | |
| 122930 | |
| CC(C)(C)C1=CC=C2C(=O)OC(=O)C2=C1 |
| C12H12O3 | |
| MFCD00060127 | |
| 4-tert-butylphthalic anhydride, 5-tert-butyl isobenzofuran-1,3-dione, 1,3-isobenzofurandione, 5-1,1-dimethylethyl, 4-t-butylphthalic anhydride, acmc-1ao20, 4-t-butyl phthalic anhydride, 4-t-butyl-phthalic anhydride, 4-tert-butylphthalicanhydride, 5-tert-butyl-2-benzo b furan-1,3-dione | |
| 5-tert-butyl-1,3-dihydro-2-benzofuran-1,3-dione |
Specifications
| 32703-79-0 | |
| White-Yellow | |
| C12H12O3 | |
| 5 g | |
| YLJYVKLZVHWUCT-UHFFFAOYSA-N | |
| 5-tert-butyl-1,3-dihydro-2-benzofuran-1,3-dione | |
| 122930 | |
| ≥98.0% (GC,T) | |
| 4-tert-Butylphthalic Anhydride |
| 78°C | |
| 136°C | |
| MFCD00060127 | |
| 4-tert-butylphthalic anhydride, 5-tert-butyl isobenzofuran-1,3-dione, 1,3-isobenzofurandione, 5-1,1-dimethylethyl, 4-t-butylphthalic anhydride, acmc-1ao20, 4-t-butyl phthalic anhydride, 4-t-butyl-phthalic anhydride, 4-tert-butylphthalicanhydride, 5-tert-butyl-2-benzo b furan-1,3-dione | |
| CC(C)(C)C1=CC=C2C(=O)OC(=O)C2=C1 | |
| 204.23 | |
| 204.23 | |
| Crystalline Powder |
Safety and Handling
TSCA : No