accessibility menu, dialog, popup

UserName

CAS RN 32703-79-0

IUPAC Name: 5-tert-butyl-1,3-dihydro-2-benzofuran-1,3-dione
Synonyms: 5-tert-butyl-2-benzofuran-1,3-dione
Molecular Weight (g/mol): 204.23
Molecular Formula: C12H12O3
InChi Key: YLJYVKLZVHWUCT-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C2C(=O)OC(=O)C2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results