missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylphthalic Anhydride 98.0+%, TCI America™

Catalog No. B13375G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B13375G 5 g
B133725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B13375G Supplier TCI America Supplier No. B13375G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 32703-79-0
Molecular Formula C12H12O3
Molecular Weight (g/mol) 204.23
MDL Number MFCD00060127
InChI Key YLJYVKLZVHWUCT-UHFFFAOYSA-N
Synonym 4-tert-butylphthalic anhydride, 5-tert-butyl isobenzofuran-1,3-dione, 1,3-isobenzofurandione, 5-1,1-dimethylethyl, 4-t-butylphthalic anhydride, acmc-1ao20, 4-t-butyl phthalic anhydride, 4-t-butyl-phthalic anhydride, 4-tert-butylphthalicanhydride, 5-tert-butyl-2-benzo b furan-1,3-dione
PubChem CID 122930
IUPAC Name 5-tert-butyl-1,3-dihydro-2-benzofuran-1,3-dione
SMILES CC(C)(C)C1=CC=C2C(=O)OC(=O)C2=C1

Specifications

Melting Point 78°C
Color White-Yellow
Boiling Point 136°C
Quantity 5 g
Formula Weight 204.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-tert-Butylphthalic Anhydride