missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-tert-Butylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$42.87 - $42.87
Specifications
| CAS | 123324-71-0 |
|---|---|
| Melting Point | 184°C |
| Molecular Formula | C10H15BO2 |
| MDL Number | MFCD01009697 |
| InChI Key | MNJYZNVROSZZQC-UHFFFAOYSA-N |
Specifications
| 123324-71-0 | |
| C10H15BO2 | |
| MNJYZNVROSZZQC-UHFFFAOYSA-N | |
| 2734320 | |
| Crystalline Powder |
| 184°C | |
| MFCD01009697 | |
| (4-tert-butylphenyl)boronic acid | |
| 178.04 |