missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. B22515G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B22515G Supplier TCI America Supplier No. B22515G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 4-tert-Butylphenylboronic Acid (contains varying amounts of Anhydride)
CAS 123324-71-0
Melting Point 184°C
Molecular Formula C10H15BO2
Quantity 5 g
MDL Number MFCD01009697
Synonym 4-t-butylphenylboronic acid, 4-tert-butylbenzeneboronic acid, 4-tert-butyl phenyl boronic acid, 4-tert-butylphenyl boronic acid, 4-tert-butyl phenylboronic acid, 4-tert-butyl-phenyl boronic acid, 4-tert-butylphenyl boranediol, p-t-butylphenylboronic acid, 4-tertbutylphenylboronic acid
InChI Key MNJYZNVROSZZQC-UHFFFAOYSA-N
SMILES CC(C)(C)C1=CC=C(C=C1)B(O)O
IUPAC Name (4-tert-butylphenyl)boronic acid
Molecular Weight (g/mol) 178.04
PubChem CID 2734320
Formula Weight 178.04
Physical Form Crystalline Powder
Show More Show Less