Learn More
4-tert-Butylphenylboronic acid, 97%
CAS: 123324-71-0 | C10H15BO2 | 178.04 g/mol
$151.28 - $151.28
Chemical Identifiers
| CAS | 123324-71-0 |
|---|---|
| Molecular Formula | C10H15BO2 |
| Molecular Weight (g/mol) | 178.04 |
| MDL Number | MFCD01009697 |
| InChI Key | MNJYZNVROSZZQC-UHFFFAOYSA-N |
| Synonym | 4-t-butylphenylboronic acid, 4-tert-butylbenzeneboronic acid, 4-tert-butyl phenyl boronic acid, 4-tert-butylphenyl boronic acid, 4-tert-butyl phenylboronic acid, 4-tert-butyl-phenyl boronic acid, 4-tert-butylphenyl boranediol, p-t-butylphenylboronic acid, 4-tertbutylphenylboronic acid |
| PubChem CID | 2734320 |
| IUPAC Name | (4-tert-butylphenyl)boronic acid |
| SMILES | CC(C)(C)C1=CC=C(C=C1)B(O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC358970050
|
Thermo Scientific Chemicals
358970050 |
5 g | Glass bottle |
Each for $151.28
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 123324-71-0 | |
| 178.04 | |
| MNJYZNVROSZZQC-UHFFFAOYSA-N | |
| 2734320 | |
| CC(C)(C)C1=CC=C(C=C1)B(O)O |
| C10H15BO2 | |
| MFCD01009697 | |
| 4-t-butylphenylboronic acid, 4-tert-butylbenzeneboronic acid, 4-tert-butyl phenyl boronic acid, 4-tert-butylphenyl boronic acid, 4-tert-butyl phenylboronic acid, 4-tert-butyl-phenyl boronic acid, 4-tert-butylphenyl boranediol, p-t-butylphenylboronic acid, 4-tertbutylphenylboronic acid | |
| (4-tert-butylphenyl)boronic acid |
Specifications
| 123324-71-0 | |
| 100.0 | |
| White | |
| 96% min. (HPLC) | |
| C10H15BO2 | |
| MFCD01009697 | |
| 4-t-butylphenylboronic acid, 4-tert-butylbenzeneboronic acid, 4-tert-butyl phenyl boronic acid, 4-tert-butylphenyl boronic acid, 4-tert-butyl phenylboronic acid, 4-tert-butyl-phenyl boronic acid, 4-tert-butylphenyl boranediol, p-t-butylphenylboronic acid, 4-tertbutylphenylboronic acid | |
| MNJYZNVROSZZQC-UHFFFAOYSA-N | |
| (4-tert-butylphenyl)boronic acid | |
| 2734320 | |
| 97% | |
| 4-tert-Butylphenylboronic acid |
| 96.0 | |
| 191.0°C to 196.0°C | |
| Authentic | |
| Glass bottle | |
| (CH3)3CC6H4B(OH)2 | |
| 5 g | |
| Solubility in water: insoluble. Other solubilities: soluble in ether,thf,dmso,dmf,methanol | |
| CC(C)(C)C1=CC=C(C=C1)B(O)O | |
| 178.04 | |
| 178.04 | |
| Crystalline Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
RUO – Research Use Only