missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylphenyl Salicylate 98.0+%, TCI America™

Catalog No. S027425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S027425G 25 g
S0274500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S027425G Supplier TCI America Supplier No. S027425G

Chemical Identifiers

CAS 87-18-3
Molecular Formula C17H18O3
Molecular Weight (g/mol) 270.328
MDL Number MFCD00045759
InChI Key DBOSBRHMHBENLP-UHFFFAOYSA-N
Synonym Salicylic Acid 4-tert-Butylphenyl Ester
PubChem CID 66597
IUPAC Name (4-tert-butylphenyl) 2-hydroxybenzoate
SMILES CC(C)(C)C1=CC=C(C=C1)OC(=O)C2=CC=CC=C2O

Specifications

Melting Point 63°C
Color White
Quantity 25 g
Formula Weight 270.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-tert-Butylphenyl Salicylate
RTECSNumber VO2020000
TSCA Yes