Learn More
4-tert-Butylphenoxyacetic acid, 98%
CAS: 1798-04-5 | C12H16O3 | 208.257 g/mol
Supplier: Thermo Scientific Chemicals A1701714
Description
Applied as a reagent for the TAC protective group in nucleosides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsApplied as a reagent for the TAC protective group in nucleosides.
Solubility
Sparingly Soluble (0.56 g/L) (25°C), Calc.
Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
Specifications
| 4-tert-Butylphenoxyacetic acid | |
| C12H16O3 | |
| 25 g | |
| Sparingly Soluble (0.56g/L) (25°C),Calc. | |
| CC(C)(C)C1=CC=C(C=C1)OCC(=O)O | |
| 208.257 | |
| 208.26 |
| 1798-04-5 | |
| MFCD00021758 | |
| 4-tert-butylphenoxyacetic acid, 4-tert-butylphenoxy acetic acid, 4-tert-butyl-phenoxy-acetic acid, 2-4-tert-butylphenoxy acetic acid, p-tert-butylphenoxy acetic acid, p-t-butylphenoxyacetic acid, 2-4-tert-butyl phenoxy acetic acid, acetic acid, 4-1,1-dimethylethyl phenoxy, acetic acid, 2-4-1,1-dimethylethyl phenoxy | |
| FBIGAJNVRFKBJL-UHFFFAOYSA-N | |
| 2-(4-tert-butylphenoxy)acetic acid | |
| 15718 | |
| 98% |
Chemical Identifiers
| 1798-04-5 | |
| 208.257 | |
| FBIGAJNVRFKBJL-UHFFFAOYSA-N | |
| 15718 | |
| CC(C)(C)C1=CC=C(C=C1)OCC(=O)O |
| C12H16O3 | |
| MFCD00021758 | |
| 4-tert-butylphenoxyacetic acid, 4-tert-butylphenoxy acetic acid, 4-tert-butyl-phenoxy-acetic acid, 2-4-tert-butylphenoxy acetic acid, p-tert-butylphenoxy acetic acid, p-t-butylphenoxyacetic acid, 2-4-tert-butyl phenoxy acetic acid, acetic acid, 4-1,1-dimethylethyl phenoxy, acetic acid, 2-4-1,1-dimethylethyl phenoxy | |
| 2-(4-tert-butylphenoxy)acetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only