missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylphenol, 97%

Catalog No. AC108000050
Change view
Click to view available options
Quantity:
5 g
1 kg
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC108000050 5 g
AC108000010 1 kg
AC108000025 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC108000050 Supplier Thermo Scientific Chemicals Supplier No. 108000050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 98-54-4 | C10H14O | 150.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 98-54-4
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
MDL Number MFCD00002367
InChI Key QHPQWRBYOIRBIT-UHFFFAOYSA-N
Synonym 4-tert-butyl phenol, butylphen, p-tert-butylphenol, ptbp, 4-1,1-dimethylethyl phenol, 4-t-butylphenol, p-t-butyl phenol, phenol, 4-1,1-dimethylethyl, p-t-butylphenol, p-terc.butylfenol
PubChem CID 7393
ChEBI CHEBI:34444
IUPAC Name 4-tert-butylphenol
SMILES CC(C)(C)C1=CC=C(C=C1)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 96.0°C to 100.0°C
Color White to Beige
Density 0.9000g/mL
Boiling Point 236.0°C to 238.0°C
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4787
Linear Formula (CH3)3CC6H4OH
Quantity 5 g
Beilstein 06, 524
Specific Gravity 0.9
Merck Index 15, 1588
Solubility Information Solubility in water: 0.6g/L (20°C). Other solubilities: soluble in aq. alcoholic alkali,soluble in alcohol and ether
Viscosity 2.4 mPa.s (110°C)
Formula Weight 150.22
Percent Purity 97%
Physical Form Flakes or Pastilles
Chemical Name or Material 4-tert-Butylphenol, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye damage.
Suspected of damaging fertility.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several
EINECSNumber 202-679-0
RTECSNumber SJ8925000
TSCA TSCA

RUO – Research Use Only